Inchi Code
1S/C11H14O/c1-4-11(12)10-6-5-8(2)7-9(10)3/h5-7H,4H2,1-3H3
Inchi Key
UWFRVQVNYNPBEF-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Liquid
Still not finding the right product?
Explore all of our products under 1-(2,4-Dimethylphenyl)propan-1-one
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
