Skip to Content
Merck

BDPH9BCCFE6B

Bide Pharmatech

4,4'-Dihydroxy-[1,1'-biphenyl]-2-carboxylic acid

Synonym(s):

4,4'-dihydroxy[1,1'-biphenyl]-2-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H10O4
CAS Number:
Molecular Weight:
230.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C13H10O4/c14-9-3-1-8(2-4-9)11-6-5-10(15)7-12(11)13(16)17/h1-7,14-15H,(H,16,17)

Inchi Key

BBGRLYLKUDNCGG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library