Skip to Content
Merck

BDPH9BCD09F1

Bide Pharmatech

2,5-Bis(trifluoromethyl)terephthalic acid

Synonym(s):

2,5-bis(trifluoromethyl)terephthalic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H4O4F6
CAS Number:
Molecular Weight:
302.13
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H4F6O4/c11-9(12,13)5-1-3(7(17)18)6(10(14,15)16)2-4(5)8(19)20/h1-2H,(H,17,18)(H,19,20)

Inchi Key

YIAGLTIEICXRQF-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library