Skip to Content

BDPH9BCD0FAE

Bide Pharmatech

1-(3,5-Difluorophenyl)propan-2-one

Synonym(s):

1-(3,5-difluorophenyl)acetone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H8O1F2
CAS Number:
Molecular Weight:
170.16
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H8F2O/c1-6(12)2-7-3-8(10)5-9(11)4-7/h3-5H,2H2,1H3

Inchi Key

GANUXJOOWCWFLI-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under 1-(3,5-Difluorophenyl)propan-2-one




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library