Skip to Content

BDPH9BCD260F

Bide Pharmatech

5-Bromo-1,2,3,4-tetrahydronaphthalen-1-ol

Synonym(s):

5-bromo-1,2,3,4-tetrahydro-1-naphthalenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H11O1Br1
CAS Number:
Molecular Weight:
227.1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H11BrO/c11-9-5-1-4-8-7(9)3-2-6-10(8)12/h1,4-5,10,12H,2-3,6H2

Inchi Key

JEDJGVSGTZKGFJ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

White to Yellow Solid



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library