Skip to Content
Merck

BDPH9BCD26B4

Bide Pharmatech

Methyl 6-bromo-1-oxo-2,3-dihydro-1H-indene-2-carboxylate

Synonym(s):

methyl 6-bromo-1-oxo-2,3-dihydro-1H-indene-2-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H9O3Br1
CAS Number:
Molecular Weight:
269.09
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H9BrO3/c1-15-11(14)9-4-6-2-3-7(12)5-8(6)10(9)13/h2-3,5,9H,4H2,1H3

Inchi Key

HQVLQXSFYKRSER-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library