Skip to Content
Merck

BDPH9BCD2CD5

Bide Pharmatech

(2-(Trifluoromethyl)-1h-indol-5-yl)methanamine

Synonym(s):

[2-(trifluoromethyl)-1H-indol-5-yl]methanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H9N2F3
CAS Number:
Molecular Weight:
214.19
MDL number:
Product BDPH9BCD2CD5 is not currently sold in your country.


Inchi Code

1S/C10H9F3N2/c11-10(12,13)9-4-7-3-6(5-14)1-2-8(7)15-9/h1-4,15H,5,14H2

Inchi Key

NBNIQGRWWWXPAD-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library