Skip to Content

BDPH9BCD2E5A

Bide Pharmatech

N-Ethyl-3,4-dimethylbenzylamine

Synonym(s):

N-(3,4-dimethylbenzyl)ethanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H17N1
CAS Number:
Molecular Weight:
163.26
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H17N/c1-4-12-8-11-6-5-9(2)10(3)7-11/h5-7,12H,4,8H2,1-3H3

Inchi Key

JTHLCDILGMWAOG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

96%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under N-Ethyl-3,4-dimethylbenzylamine



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library