Skip to Content
Merck

BDPH9BCD3032

Bide Pharmatech

2-(Chloromethyl)-1,4-difluorobenzene

Synonym(s):

2-(chloromethyl)-1,4-difluorobenzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H5Cl1F2
CAS Number:
Molecular Weight:
162.57
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H5ClF2/c8-4-5-3-6(9)1-2-7(5)10/h1-3H,4H2

Inchi Key

INXKTZMJFPRVAY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library