Skip to Content
Merck

BDPH9BCD3553

Bide Pharmatech

2-Bromo-1,3-bis(3-methoxyphenyl)propane-1,3-dione

Synonym(s):

2-bromo-1,3-bis(3-methoxyphenyl)-1,3-propanedione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C17H15O4Br1
CAS Number:
Molecular Weight:
363.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C17H15BrO4/c1-21-13-7-3-5-11(9-13)16(19)15(18)17(20)12-6-4-8-14(10-12)22-2/h3-10,15H,1-2H3

Inchi Key

TUECNKMLFJLFOE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid or viscous liquid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library