Skip to Content
Merck

BDPH9BCD4B7C

Bide Pharmatech

3-Fluoro-5-methylpyridine

Synonym(s):

3-fluoro-5-methylpyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H6N1F1
CAS Number:
Molecular Weight:
111.12
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H6FN/c1-5-2-6(7)4-8-3-5/h2-4H,1H3

Inchi Key

CKQXPRSCSFUGRC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Colorless to Pale-yellow to Yellow-brown Liquid



Still not finding the right product?

Explore all of our products under 3-Fluoro-5-methylpyridine


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library