Skip to Content
Merck

BDPH9BCD4E2E

Bide Pharmatech

4-Chloro-6-(trifluoromethyl)nicotinaldehyde

Synonym(s):

4-chloro-6-(trifluoromethyl)nicotinaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H3O1N1Cl1F3
CAS Number:
Molecular Weight:
209.55
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H3ClF3NO/c8-5-1-6(7(9,10)11)12-2-4(5)3-13/h1-3H

Inchi Key

FBPZVYHRKMIAKN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library