Skip to Content
Merck

BDPH9BCD6568

Bide Pharmatech

2,6-Dibromo-4-(trifluoromethyl)phenol

Synonym(s):

2,6-dibromo-4-(trifluoromethyl)phenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H3O1Br2F3
CAS Number:
Molecular Weight:
319.9
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H3Br2F3O/c8-4-1-3(7(10,11)12)2-5(9)6(4)13/h1-2,13H

Inchi Key

IEMNQLAXBXXIKT-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid or lump



Still not finding the right product?


hstatements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library