Skip to Content

BDPH9BCD72FD

Bide Pharmatech

2-Bromo-5-chloro-3-(trifluoromethyl)pyridine

Synonym(s):

2-bromo-5-chloro-3-(trifluoromethyl)pyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H2N1Cl1Br1F3
CAS Number:
Molecular Weight:
260.44
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H2BrClF3N/c7-5-4(6(9,10)11)1-3(8)2-12-5/h1-2H

Inchi Key

GDQVCXJOIDBNRP-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid or liquid



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library