Skip to Content
Merck

BDPH9BCD898B

Bide Pharmatech

2-Chloro-7-methyl-7H-purin-6-ol

Synonym(s):

2-chloro-7-methyl-1,7-dihydro-6H-purin-6-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5O1N4Cl1
CAS Number:
Molecular Weight:
184.58
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H5ClN4O/c1-11-2-8-4-3(11)5(12)10-6(7)9-4/h2H,1H3,(H,9,10,12)

Inchi Key

HTRDGUDBXFNIOU-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 2-Chloro-7-methyl-7H-purin-6-ol


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library