Skip to Content
Merck

BDPH9BCD966F

Bide Pharmatech

3-Iodo-5-methoxy-1H-pyrrolo[2,3-b]pyridine

Synonym(s):

3-iodo-1H-pyrrolo[2,3-b]pyridin-5-yl methyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H7O1N2I1
Molecular Weight:
274.06
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H7IN2O/c1-12-5-2-6-7(9)4-11-8(6)10-3-5/h2-4H,1H3,(H,10,11)

Inchi Key

FKIMIWSUCKEXLD-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library