Skip to Content
Merck

BDPH9BCDA1E9

Bide Pharmatech

1-(2-Bromophenyl)propan-2-ol

Synonym(s):

1-(2-bromophenyl)-2-propanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H11O1Br1
CAS Number:
Molecular Weight:
215.09
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H11BrO/c1-7(11)6-8-4-2-3-5-9(8)10/h2-5,7,11H,6H2,1H3

Inchi Key

RJKPOWPTZRARKB-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Liquid or semi-solid or solid



Still not finding the right product?

Explore all of our products under 1-(2-Bromophenyl)propan-2-ol




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library