Skip to Content
Merck

BDPH9BCDA656

Bide Pharmatech

3,4,5-Trifluorobenzenesulfonamide

Synonym(s):

3,4,5-trifluorobenzenesulfonamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H4O2N1S1F3
CAS Number:
Molecular Weight:
211.16
MDL number:
Product BDPH9BCDA656 is not currently sold in your country.


Inchi Code

1S/C6H4F3NO2S/c7-4-1-3(13(10,11)12)2-5(8)6(4)9/h1-2H,(H2,10,11,12)

Inchi Key

BIAGBHUIRSMWRI-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 3,4,5-Trifluorobenzenesulfonamide



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library