Skip to Content
Merck

BDPH9BCDAAF3

Bide Pharmatech

6,7-Dichloro-1H-indole-2-carboxylic acid

Synonym(s):

6,7-dichloro-1H-indole-2-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H5O2N1Cl2
CAS Number:
Molecular Weight:
230.05
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H5Cl2NO2/c10-5-2-1-4-3-6(9(13)14)12-8(4)7(5)11/h1-3,12H,(H,13,14)

Inchi Key

TUXCSCQQWQOWDU-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library