Skip to Content

BDPH9BCDB80F

Bide Pharmatech

(5-Bromo-1H-indol-2-yl)methanol

Synonym(s):

(5-bromo-1H-indol-2-yl)methanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H8O1N1Br1
CAS Number:
Molecular Weight:
226.07
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H8BrNO/c10-7-1-2-9-6(3-7)4-8(5-12)11-9/h1-4,11-12H,5H2

Inchi Key

ZIUDOZPLPNFTMR-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Form: solid



Still not finding the right product?

Explore all of our products under (5-Bromo-1H-indol-2-yl)methanol


hstatements

Precautionary Statements

Signal Word

Danger

Pictograms

Corrosion


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library