Skip to Content
Merck

BDPH9BCDCCFB

Bide Pharmatech

3-(Isopropyl(methyl)amino)propanethioamide

Synonym(s):

3-[isopropyl(methyl)amino]propanethioamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H16N2S1
CAS Number:
Molecular Weight:
160.28
MDL number:
Product BDPH9BCDCCFB is not currently sold in your country.


Inchi Code

1S/C7H16N2S/c1-6(2)9(3)5-4-7(8)10/h6H,4-5H2,1-3H3,(H2,8,10)

Inchi Key

YIQVOFAZZREHCK-UHFFFAOYSA-N

Shipping Temperature

Cold-chain transportation

Purity

95%

Storage temp.

Sealed in dry, store in freezer, under -20C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library