Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C6H6BrClN2/c7-3-1-2-4(8)6(10)5(3)9/h1-2H,9-10H2
Inchi Key
CMKDPCHKJRWRKB-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Keep in dark place, inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 3-Bromo-6-chlorobenzene-1,2-diamine
hstatements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
