Skip to Content
Merck

BDPH9BCDDA67

Bide Pharmatech

4,7-Dibromo-1H-benzo[d]imidazole

Synonym(s):

4,7-dibromo-1H-benzimidazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H4N2Br2
CAS Number:
Molecular Weight:
275.93
MDL number:
Product BDPH9BCDDA67 is not currently sold in your country.


Inchi Code

1S/C7H4Br2N2/c8-4-1-2-5(9)7-6(4)10-3-11-7/h1-3H,(H,10,11)

Inchi Key

ZGLGYSCEKIFGMB-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

Yellow to Brown Solid

Storage temp.

Keep in dark place, sealed in dry, 2-8°C



Still not finding the right product?

Explore all of our products under 4,7-Dibromo-1H-benzo[d]imidazole




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library