Skip to Content
Merck

BDPH9BCDDE03

Bide Pharmatech

1-Acetyl-3-bromoindole

Synonym(s):

1-acetyl-3-bromo-1H-indole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H8O1N1Br1
CAS Number:
Molecular Weight:
238.08
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H8BrNO/c1-7(13)12-6-9(11)8-4-2-3-5-10(8)12/h2-6H,1H3

Inchi Key

AYFPZXVRXRMCQX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Yellow to Brown Solid



Still not finding the right product?

Explore all of our products under 1-Acetyl-3-bromoindole


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library