Skip to Content

BDPH9BCDE0EC

Bide Pharmatech

Methyl 4-amino-3-formylbenzoate

Synonym(s):

methyl 4-amino-3-formylbenzoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H9O3N1
CAS Number:
Molecular Weight:
179.18
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H9NO3/c1-13-9(12)6-2-3-8(10)7(4-6)5-11/h2-5H,10H2,1H3

Inchi Key

VNDSSJJSVMYZKF-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Solid



Still not finding the right product?

Explore all of our products under Methyl 4-amino-3-formylbenzoate


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library