Skip to Content
Merck

BDPH9BCDE43E

Bide Pharmatech

(2-Cyano-3-fluorophenyl)boronic acid

Synonym(s):

2-cyano-3-fluorophenylboronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H5O2N1F1B1
CAS Number:
Molecular Weight:
164.93
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H5BFNO2/c9-7-3-1-2-6(8(11)12)5(7)4-10/h1-3,11-12H

Inchi Key

SJGAIKIHINNTDX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library