Skip to Content

BDPH9BCDEB28

Bide Pharmatech

3-Methoxy-4-(methylsulfonyl)aniline

Synonym(s):

3-methoxy-4-(methylsulfonyl)aniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H11O3N1S1
CAS Number:
Molecular Weight:
201.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H11NO3S/c1-12-7-5-6(9)3-4-8(7)13(2,10)11/h3-5H,9H2,1-2H3

Inchi Key

LAYYXXQNPGIKED-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library