Skip to Content
Merck

BDPH9BCDF1D4

Bide Pharmatech

1H-Indole-2,5-dicarboxylic acid

Synonym(s):

1H-indole-2,5-dicarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H7O4N1
CAS Number:
Molecular Weight:
205.17
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H7NO4/c12-9(13)5-1-2-7-6(3-5)4-8(11-7)10(14)15/h1-4,11H,(H,12,13)(H,14,15)

Inchi Key

MUABQYJAPOQWCH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Solid

Storage temp.

Sealed in dry, 2-8C



Still not finding the right product?

Explore all of our products under 1H-Indole-2,5-dicarboxylic acid


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library