Skip to Content
Merck

BDPH9BCDF327

Bide Pharmatech

3-Bromo-2-chloro-6-fluorobenzoic acid

Synonym(s):

3-bromo-2-chloro-6-fluorobenzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H3O2Cl1Br1F1
CAS Number:
Molecular Weight:
253.45
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H3BrClFO2/c8-3-1-2-4(10)5(6(3)9)7(11)12/h1-2H,(H,11,12)

Inchi Key

BPKBUPFQEALBFX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library