Skip to Content

BDPH9BCE221D

Bide Pharmatech

tert-Butyl (6-chloro-4-iodopyridin-3-yl)carbamate

Synonym(s):

tert-butyl 6-chloro-4-iodo-3-pyridinylcarbamate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H12O2N2Cl1I1
CAS Number:
Molecular Weight:
354.57
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H12ClIN2O2/c1-10(2,3)16-9(15)14-7-5-13-8(11)4-6(7)12/h4-5H,1-3H3,(H,14,15)

Inchi Key

YCBOUHLCYYKTFG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

White to Pale-yellow to Yellow-brown Solid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library