Skip to Content
Merck

BDPH9C01EC71

Bide Pharmatech

Tyr-Tyr-Tyr-Tyr-Tyr-Tyr

Synonym(s):

(2S,5S,8S,11S,14S,17S)-17-amino-2,5,8,11,14-pentakis(4-hydroxybenzyl)-18-(4-hydroxyphenyl)-4,7,10,13,16-pentaoxo-3,6,9,12,15-pentaazaoctadecan-1-oic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C54H56O13N6
CAS Number:
Molecular Weight:
997.07
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C54H56N6O13/c55-43(25-31-1-13-37(61)14-2-31)49(67)56-44(26-32-3-15-38(62)16-4-32)50(68)57-45(27-33-5-17-39(63)18-6-33)51(69)58-46(28-34-7-19-40(64)20-8-34)52(70)59-47(29-35-9-21-41(65)22-10-35)53(71)60-48(54(72)73)30-36-11-23-42(66)24-12-36/h1-24,43-48,61-66H,25-30,55H2,(H,56,67)(H,57,68)(H,58,69)(H,59,70)(H,60,71)(H,72,73)/t43-,44-,45-,46-,47-,48-/m0/s1

Inchi Key

JUUZKZKBBSFAAT-RYICAFHUSA-N

Shipping Temperature

Cold-chain transportation

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Powder or crystals




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library