Skip to Content
Merck

BDPH9C01FE29

Bide Pharmatech

1-(3-Chloropropyl)azepane hydrochloride

Synonym(s):

1-(3-chloropropyl)azepane hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H19N1Cl2
CAS Number:
Molecular Weight:
212.16
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H18ClN.ClH/c10-6-5-9-11-7-3-1-2-4-8-11;/h1-9H2;1H

Inchi Key

GQEZLLGQUIRVHL-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library