Skip to Content
Merck

BDPH9C020EFA

Bide Pharmatech

(3-(Benzyloxy)prop-1-en-2-yl)benzene

Synonym(s):

(3-(benzyloxy)prop-1-en-2-yl)benzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C16H16O1
CAS Number:
Molecular Weight:
224.3
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C16H16O/c1-14(16-10-6-3-7-11-16)12-17-13-15-8-4-2-5-9-15/h2-11H,1,12-13H2

Inchi Key

ZMDWEVRZQXRVDU-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library