Skip to Content
Merck

BDPH9C021E04

Bide Pharmatech

(Iminomethyl)-L-glutamic acid

Synonym(s):

(2S)-2-[(iminomethyl)amino]pentanedioic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H10O4N2
CAS Number:
Molecular Weight:
174.16
MDL number:
Product BDPH9C021E04 is not currently sold in your country.


Inchi Code

1S/C6H10N2O4/c7-3-8-4(6(11)12)1-2-5(9)10/h3-4H,1-2H2,(H2,7,8)(H,9,10)(H,11,12)/t4-/m0/s1

Inchi Key

NRXIKWMTVXPVEF-BYPYZUCNSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under (Iminomethyl)-L-glutamic acid



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library