Skip to Content

BDPH9C021F01

Bide Pharmatech

(1S,3S)-Methyl 3-hydroxycyclohexanecarboxylate

Synonym(s):

methyl (1S,3S)-3-hydroxycyclohexane-1-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H14O3
CAS Number:
Molecular Weight:
158.2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H14O3/c1-11-8(10)6-3-2-4-7(9)5-6/h6-7,9H,2-5H2,1H3/t6-,7-/m0/s1

Inchi Key

OXQRLBFDJMSRMM-BQBZGAKWSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library