Skip to Content
Merck

BDPH9C02488F

Bide Pharmatech

5-(Furan-2-yl)-1H-pyrazol-3-amine

Synonym(s):

5-(2-furyl)-1H-pyrazol-3-ylamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H7O1N3
CAS Number:
Molecular Weight:
149.15
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H7N3O/c8-7-4-5(9-10-7)6-2-1-3-11-6/h1-4H,(H3,8,9,10)

Inchi Key

XJNZHICOWTVWOX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

White to light-brown to brown powder or crystals



Still not finding the right product?

Explore all of our products under 5-(Furan-2-yl)-1H-pyrazol-3-amine


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library