Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C19H27D5O2
Molecular Weight:
297.49
UNSPSC Code:
41116107
agency
NMI Australia certified reference material
manufacturer/tradename
NMI Australia
format
neat
storage temp.
2-8°C
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Carc. 2 - Repr. 1B
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
