Skip to Content
Merck

ENA432540435

Enamine

2-(1-methyl-1H-indazol-3-yl)acetic acid

Sign In to View Organizational & Contract Pricing

Select a Size

Product ENA432540435 is not currently sold in your country.

About This Item

CAS Number:
Molecular Weight:
190.2
MDL number:
Product ENA432540435 is not currently sold in your country.

Iupac Name

(1-methyl-1H-indazol-3-yl)acetic acid

Inchi Code

1S/C10H10N2O2/c1-12-9-5-3-2-4-7(9)8(11-12)6-10(13)14/h2-5H,6H2,1H3,(H,13,14)

InChI key

YEXWKCLXINFBDI-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-199330

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-199330


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library