Skip to Content
Merck

1.17884

Magnetic perforated lid Microbial Air Sampler

new

for use with MAS-100 Sirius with 90 mm settle plates at 200 SLPM flow rate

Sign In to View Organizational & Contract Pricing

About This Item

UNSPSC Code:
41103403
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

material

stainless steel lid

product line

MAS-100®

feature

no. of holes 400 X 0.7 mm

packaging

pkg of 1 unit

manufacturer/tradename

(Perforated Lid Sirius Type B 90 mm)

parameter

200 L/min flow rate

application(s)

air monitoring

compatibility

for use with MAS-100® Sirius

Looking for similar products? Visit Product Comparison Guide

General description

Perforated lids are crucial for optimizing the physical and biological sampling efficiency of Andersen impaction-based microbial air samplers. Air is accelerated through the perforations in the lid, creating a jet that impacts the agar surface.
The Lid for MAS-100 Sirius Microbial Air Samplers is designed to fit tightly on the instrument by easy and quick magnetic closure without twisting.
It is made from 316L stainless-steel which has good resistance towards a broad range of disinfectants including vaporized hydrogen peroxide and is suitable for autoclaving.
The magnetic lid type B 90 mm has a sieve geometry of 400 holes with a diameter of 0.7 mm and is suitable for use with 90 mm agar plates. This sieve configuration delivers a nominal d50 value of 1.15 µm at a flow rate of 200 SLPM (standard liter per minute).
Each lid is marked with laser-based marking without significantly altering the properties of the metal surface. The information of the unique marking includes the identification code of the perforated lid as well as the serial number in barcode and readable format.

Application

The perforated lid is designed for active air monitoring with MAS-100 Sirius instruments at a flow rate of 200 standard Liter per minute (SLPM) using 90 mm agar plates such as ICR/ICRplus settle plates, enabling efficient air sampling in Isolators and cleanrooms.

Features and Benefits

  • Independently validated efficiency acc. ISO 14698 and EN 17141 at a flow rate of 200 SLPM with ICR settle plates
  • Stainless-steel for optimized cleaning and sterilization
  • Unique marking allows manual and digital tracking

Legal Information

MAS-100 is a registered trademark of MBV AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Discover the MAS-100 Sirius® Microbial Air Sampler, designed for reliable air monitoring. Explore features, training, and support services to enhance your lab's compliance and efficiency.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service