Skip to Content
Merck

MB1HC10FS36AS

Millistak+® HC Pod Depth Filter

new

B1HC, filtration area 1.1 m2, Process Scale in APAC format Optimized Packaging 36/PK

Synonym(s):

1.1m2 surface aera, Millistak+® HC Pod Depth Filter, Optimized Packaging, Process Scale Pod

Sign In to View Organizational & Contract Pricing

Select a Size


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

material

acetal (polypropylene)
mixed cellulose esters (MCE) filter aid

Quality Level

product line

Millistak+®

feature

hydrophilic

filtration area

1.1 m2

impurities

(LAL test)

input

liquid

fitting

(Flat Seal)

application(s)

pharma/biopharma processes

code

B1HC

shipped in

ambient

General description

To improve sustainability of the packaging and shipping of filter products, Optimized Pack solutions for Millistak+® HC Process Scale Pod filters have been developed to optimize transport and receiving processes as well as to reduce waste.

Application

Post TFF Clarification fluids

Features and Benefits

The key benefits are:
  • 24% reduction in corrugated packaging waste to recycle or dispose.
  • 94% reduction of paper documentation.
  • 12% decrease in number of deliveries which will further reduce energy use and emissions.
  • 70% reduction in operator time to open and manage the product and packaging.

Packaging

36/pack Optimized packaging configuration includes 36 pods distributed in 3 boxes on one APAC standardized pallet (pallet size: 1100*1100mm).

Other Notes

Integrity is maintained after 1 autoclave cycle of 60 minutes at 123 °C

Legal Information

MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service