Skip to Content
Merck

MSGLASS01

MilliSnap System protective glass

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€289.00

€289.00


Please contact Customer Service for Availability

Request more information

About This Item

UNSPSC Code:
41116500
NACRES:
NB.24

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

manufacturer/tradename

MilliSnap

Quality Level

input

pharmaceutical(s)

application(s)

bioburden testing
pharmaceutical
water monitoring

shipped in

ambient

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
MSLM10001MSSM10001MSSM25001
application(s)

bioburden testing
pharmaceutical
water monitoring

application(s)

bioburden testing
pharmaceutical
water monitoring

application(s)

bioburden testing
pharmaceutical
water monitoring

application(s)

bioburden testing
pharmaceutical
water monitoring

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

manufacturer/tradename

MilliSnap®

manufacturer/tradename

MilliSnap®

manufacturer/tradename

MilliSnap®

manufacturer/tradename

MilliSnap®

input

pharmaceutical(s)

input

-

input

-

input

-

Quality Level

400

Quality Level

400

Quality Level

400

Quality Level

400

General description

Designed to simplify your workflow, the MilliSnap plug and play system is a hands-free method for separating your Milliflex funnel and attaching the membrane to the Millipore media cassettes. The unique convex shape of the Milliflex membrane combined with the MilliSnap system ensures proper contact between membrane and media.

Legal Information

MILLISNAP is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service