Skip to Content
Merck

ZBYPASSKT

Emergency Backup Function for AFS® Essential systems

Ensures uninterrupted supply of pure water

Sign In to View Organizational & Contract Pricing

Select a Size

1 EA

€387.00

€387.00


Estimated to ship TODAYDetails


Request more information

About This Item

UNSPSC Code:
41104214
eCl@ss:
33050190
NACRES:
NA.21

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

compatibility

for use with AFS®

shipped in

ambient

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
ZLXLPTCABCP1ALLRESZLXE0030WW
compatibility

for use with AFS®

compatibility

for use with Milli-Q® CLX, for use with Milli-Q® HR, for use with Milli-Q® HX

compatibility

for use with AFS® (1999-2012)

compatibility

-

shipped in

ambient

shipped in

-

shipped in

ambient

shipped in

-

General description

Emergency Backup Function for AFS® Essential systems is an easy and quick to install water purification accessory. Used as a backup for AFS® systems, it enables the uninterrupted supply of pure water when the unit is being serviced.

Application

For uninterrupted water supply

Legal Information

AFS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service