Skip to Content
Merck

Skip To

4

Niaproof® 4

~27% in H2O

Synonym(s):

7-Ethyl-2-methyl-4-undecyl sulfate sodium salt, Sodium 7-ethyl-2-methyl-4-undecyl sulfate, Tergitol™ 4

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C14H29NaO4S
CAS Number:
Molecular Weight:
316.43
UNSPSC Code:
12161900
PubChem Substance ID:
MDL number:


description

anionic

form

liquid

concentration

~27% in H2O

SMILES string

[Na+].CCCCC(CC)CCC(CC(C)C)OS([O-])(=O)=O

InChI

1S/C14H30O4S.Na/c1-5-7-8-13(6-2)9-10-14(11-12(3)4)18-19(15,16)17;/h12-14H,5-11H2,1-4H3,(H,15,16,17);/q;+1/p-1

InChI key

FVEFRICMTUKAML-UHFFFAOYSA-M

Other Notes

Alkyl sodium sulfate surfactant.
These are the same anionic surfactants formerly produced by Union Carbide under the Tergitol name.

Legal Information

Niaproof is a registered trademark of Niacet Corp.
TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow


Still not finding the right product?

Explore all of our products under Niaproof® 4


Pictograms

Corrosion

Signal Word

Danger

Hazard Codes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters