Skip to Content
Merck

Skip To

BPS78371

PARP Substrate Mixture 2

Synonym(s):

PARP Substrate Mixture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
47101613
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

BPS Bioscience

storage temp.

-65 to -96°C

General description

Proprietary mixture including biotinylated and non-biotinylated NAD+, optimized for use with BPS Bioscience′s poly(ADP) ribose polymerase (PARP) assay kits:
  • PARP6 Chemiluminescent Assay Kit (BPS Bioscience #80556)
  • PARP7 Chemiluminescent Assay Kit (BPS Bioscience #79729)
  • PARP11 Chemiluminescent Assay Kit (BPS Bioscience #80561)
  • PARP14 Chemiluminescent Assay Kit (BPS Bioscience #80568)

Application

Suitable for use with cell extracts and purified PARP enzymes.

Datasheet and Protocols may be found here.

Preparation Note

Tris-buffered solution

Analysis Note

Avoid freeze/thaw cycles.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BPS78366493800528822
description

-

description

-

description

The PARP Inhibitor VI, NU1025, also referenced under CAS 90417-38-2, controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.

description

The PARP Inhibitor XII, also referenced under CAS 489457-67-2, controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.

storage temp.

-65 to -96°C

storage temp.

-65 to -96°C

storage temp.

−20°C

storage temp.

2-8°C

manufacturer/tradename

BPS Bioscience

manufacturer/tradename

BPS Bioscience

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®


Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library