Skip to Content
Merck

Skip To

BPS80584

PARPtrap Assay Kit for PARP1

Synonym(s):

Assay Kits, ADPRT, poly (ADP-ribose) polymerase 1, poly(ADP-ribosyl)transferase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

eCl@ss:
32160702
UNSPSC Code:
41116100
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

BPS Bioscience

technique(s)

activity assay: suitable (Fluorescence Polarization)

UniProt accession no.

shipped in

dry ice

storage temp.

-65 to -96°C

General description

The PARPtrap Assay Kit for PARP1 is a fluorescence polarization (FP), homogeneous, 96-well or 384-well assay designed for screening and profiling small molecules that trap PARP1 onto DNA and complex formation in a high throughput screening (HTS) format. The PARPtrap Assay Kit for PARP1 comes in a convenient 96-well or 384-well format, with purified recombinant PARP1 enzyme, a fluorescent-labeled oligonucleotide duplex, NAD+ and PARPtrap assay buffer for 100 or 400 enzyme reactions.

Application

Screen small molecules that enhance PARP1/DNA trapping for drug discovery and high throughput screening (HTS) applications, and IC50 determination.


PARP1, also known as poly-(ADP-ribose) polymerase 1 or NAD+ ADP-ribosyltransferase 1, is part of the PARP family, and it is the most abundant member. ADP ribosylation, which is the addition of an ADP-ribose to a protein, is a reversible post-translational modification of proteins mostly involved in the DNA Damage Response (DDR) pathway. Poly-ADP-ribosylation (termed PARylation) is the addition of linear or branched chains of ADP-ribose. PARP1 participates in DNA repair by non-homologous end joining (NHEJ), homologous recombination (HR), microhomology-mediated end-joining (MMEJ) and nucleotide excision repair. Dysfunction of DDR pathways can lead to oncogenesis. Overexpression of PARP1 has been found in breast and colon cancer, neuroblastoma, and others. This overexpression can lead to increasing MMEJ, an error-prone DNA repair mechanism, and genome instability leading to cancer. In addition to being involved in DDR, PARP1 is also linked to inflammation and type I diabetes. PARP1 is known to bind damaged DNA through its DNA-binding domains. Binding to DNA activates PARP1 and in the presence of NAD+ PARP1 ribosylates itself (auto-ribosylation), what in consequence leads to PARP1 dissociation from the DNA due to the accumulated negative charge of the ribosyl polymer. In the presence of some inhibitors, however, PARP remains bound to DNA, a phenomenon termed trapping. Trapped PARP-DNA complexes have been shown to be highly cytotoxic to cancer cells, therefore such inhibitors may be desirable for cancer treatment. Further understanding of the molecular pathways involving PARP1, and this contribution to disease, will continue to pave the way for new therapies for PARP1-linked diseases.


Datasheet and Protocols for the 96 reaction kit may be found here.

Datasheet an Protocols for the 384 reactoin kit may be found here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SRP0192HPA045168AV33754
description

-

description

recombinant, expressed in baculovirus infected insect cells, ≥80% (SDS-PAGE)

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

IgG fraction of antiserum

technique(s)

activity assay: suitable (Fluorescence Polarization)

technique(s)

inhibition assay: suitable

technique(s)

immunohistochemistry: 1:500-1:1000, immunoblotting: 0.04-0.4 μg/mL, immunofluorescence: 0.25-2 μg/mL

technique(s)

western blot: suitable

storage temp.

-65 to -96°C

storage temp.

−70°C

storage temp.

−20°C

storage temp.

−20°C

manufacturer/tradename

BPS Bioscience

manufacturer/tradename

Sigma-Aldrich

manufacturer/tradename

-

manufacturer/tradename

-

UniProt accession no.

P09874

UniProt accession no.

P09874

UniProt accession no.

P09874

UniProt accession no.

P09874

shipped in

dry ice

shipped in

dry ice

shipped in

wet ice

shipped in

-


Storage Class

10 - Combustible liquids

WGK

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library