Skip to Content
Merck

MTCD334024

MTC Bio OptiWell Tip Comb

new

24 tip, V-bottom, polypropylene, non-sterile, pack of 50 ea (25 bags of 2ea)

Synonym(s):

MTC Bio D3340-24, automation comb, automation tool, comb, extraction comb

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
1121800

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

material

V-bottom
polypropylene

sterility

non-sterile

packaging

pack of 50 ea (25 bags of 2ea)

manufacturer/tradename

MTC Bio D3340-24

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

OptiWell Tip Combs are designed for magnetic bead-based nucleic acid and protein affinity purification in automated systems including Kingfisher Flex and Accuris IsoPure extraction systems. They can be used for viral RNA extraction, DNA extraction, immunoprecipitation and quantification and are made of chemically inert, low binding virgin polypropylene. Certified RNase, DNase and pyrogen free.

Application

DNA extraction, quantification, viral RNA extraction, immunoprecipitations

Features and Benefits

  • Compatable with automated systems including Kingfisher Flex and Accuris IsoPure extraction systems.
  • Low binding virgin polypropylene.
  • Certified RNase, DNase and pyrogen free.

Legal Information

IsoPure is a trademark of Benchmark Scientific, Inc.
MTC is a trademark of MTC Bio

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service