Merck
MTCP3010AP
MTC Bio Pure.Amp™ PCR 8 Tube Strips
new
capacity 0.2 mL, flat cap (attached), purple, pack of 120 ea
Synonym(s):
MTC Bio Pure.Amp PCR 8 Tube Strips, MTC Bio P3010-A-P, PCR sample storage tubes, sample storage tube, snap closure tube
Sign In to View Organizational & Contract Pricing
Select a Size
MTCP3010AP-120EA
€105.00
€105.00
Please contact Customer Service for Availability
About This Item
UNSPSC Code:
41121800
Skip To
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
flat cap (attached)
polypropylene
purple
packaging
pack of 120 ea
manufacturer/tradename
MTC Bio P3010-A-P
capacity
0.2 mL
General description
Pure.Amp 0.2mL PCR strips can be supplied with domed caps or flat caps and can be attached or supplied separately, color-coded tubes are easily identified at a glance, making them the ideal tube to track your samples. Strips are precision manufactured to maintain tight dimensional tolerances of ultra-thin wall thickness, for improved thermal transfer and are certified free of RNase, DNase, DNA and PCR inhibitors, and are made of heat-resistant medical-grade polypropylene.
Features and Benefits
  • RNase, DNase, DNA, and PCR inhibitor free
  • Ultra-thin, uniform wall thickness
Legal Information
MTC is a trademark of MTC Bio
Pure.Amp is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)