Skip to Content
Merck

Skip To

OGS96

PSF-CMV-NH2-FLAG®-EKT-NCOI - N-TERMINAL FLAG® TAG PLASMID

plasmid vector for molecular cloning

Synonym(s):

cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.85
UNSPSC Code:
12352200
Promoter:
Promoter name: CMV
Promoter activity: constitutive
Promoter type: mammalian
Origin of replication:
pUC (500 copies)
Bacteria selection:
kanamycin
Reporter gene:
none
Peptide cleavage:
no cleavage


tag

FLAG® tagged

form

buffered aqueous solution

mol wt

size 4247 bp

technique(s)

molecular cloning: suitable

bacteria selection

kanamycin

origin of replication

pUC (500 copies)

peptide cleavage

no cleavage

peptide tag location

N-terminal

promoter

Promoter name: CMV
Promoter activity: constitutive
Promoter type: mammalian

reporter gene

none

shipped in

ambient

storage temp.

−20°C

General description

This plasmid adds a FLAG epitope tag to the N-terminus of a protein that is encoded within the multiple cloning site. This tag allows the detection and purification of a tagged protein using antibodies raised against the FLAG epitope. The FLAG tag coding sequence is DYKDDDDK. There is an enterokinase cleavage site (DDDDK) within the FLAG tag sequence that can be used to remove it from a purified protein if required. It cleaves after the lysine residue.

Promoter Expression Level: This plasmid contains the mammalian CMV promoter to drive gene expression. We have tested all of our mammalian promoters in a range of cell types and CMV is consistently the strongest in those we have studied. However there are many reports of the CMV promoter demonstrating silencing by methylation in long-term culture.

Application

Cloning in a gene: This vector has been designed to allow the addition of a peptide tag to the end of a protein of interest using standard cloning techniques.Multiple Cloning Site Notes:

There is a start codon in the NcoI site can be removed by digestion with KpnI if required. The MCS for gene insertions extends from NotI to XbaI however the tag resides between the NotI and HindIII sites. There are Shine-Dalgarno sequences and KOZAK sequences aligned with the start codon of the peptide tag.

The ClaI to NheI sites have other functions such as adding C-terminal peptide tags second promoters or IRES expression components. The BsgI and BseRI restriction sites cleave within the stop codon in the XbaI site and allow the retrospective fusion of C-terminal peptide tags sequences if the stop codon is placed in this position.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxgene.com

Other Notes

To view sequence information for this product, please visit the product page

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

12 - Non Combustible Liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters