Skip to Content
Merck

Skip To

S192

Anti-SDF-1/PBSF antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Stromal Cell-Derived Factor-1/Pre-B Cell Growth Stimulating Factor

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
51111800
MDL number:
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA (i), Neutral, WB
Citations:
8


biological source

goat

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

indirect ELISA: 0.5-1 μg/mL, neutralization: suitable, western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CXCL12(6387)

General description

Stromal cell-derived factor-1 (SDF-1) is a proinflammatory cytokine that belongs to the CXC family of chemokines. The effects of SDF-1 are mediated by its binding to the receptor CXCR4, and sometimes to CXCR7. Both lymphoid (bone marrow, spleen and thymus) and non-lymphoid (stomach, kidney, liver, lung, brain, heart) organs express SDF-1. The most important functions assigned to SDF-1 are angiogenesis, migration of μglia and astrocytes, tissue repair and as chemoattractant to hematopoietic cells. Studies also implicate SDF-1 in survival of human glioma cells via the activation of PI3K/Akt pathway. The other pathways that are activated by SDF-1 include MAPK, NF-κB, paxillin, Crk and other focal adhesion proteins. A variety of human malignancies of pancreas, breast, prostate and glioma exhibit overexpression of CXCR4. High expression of SDF-1 has been observed in lymph nodes, lungs, bone and liver that are the most common sites of metastasis of breast cancer cells
This antibody can be used for the localization and detection of human SDF-1/PBSF. It can detect human SDF-1α and SDF-1β.

Immunogen

recombinant human SDF-1/PBSF expressed in E. coli.

Application

Anti-SDF-1/PBSF antibody may be used for immunoblotting at a working concentration of 0.1-0.2 μg/ml. For ELISA, the recommended working antibody concentration is 0.5-1.0 μg/ml. The antibody is also suitable for neutralization assays.

Biochem/physiol Actions

The antibody may be used for neutralization of SDF-1α and SDF-1β, and for localization and detection of human SDF-1/PBSF.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose..

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters