Skip to Content
Merck

SAB1411045

Anti-PGM2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ10983, MSTP006

Sign In to View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 68.3 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PGM2(55276)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
HPA040676HPA060173HPA029518
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

form

buffered aqueous solution

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

Immunogen

PGM2 (NP_060760.2, 1 a.a. ~ 612 a.a) full-length human protein.

Sequence
MAAPEGSGLGEDARLDQETAQWLRWDKNSLTLEAVKRLIAEGNKEELRKCFGARMEFGTAGLRAAMGPGISRMNDLTIIQTTQGFCRYLEKQFSDLKQKGIVISFDARAHPSSGGSSRRFARLAATTFISQGIPVYLFSDITPTPFVPFTVSHLKLCAGIMITASHNPKQDNGYKVYWDNGAQIISPHDKGISQAIEENLEPWPQAWDDSLIDSSPLLHNPSASINNDYFEDLKKYCFHRSVNRETKVKFVHTSVHGVGHSFVQSAFKAFDLVPPEAVPEQKDPDPEFPTVKYPNPEEGKGVLTLSFALADKTKARIVLANDPDADRLAVAEKQDSGEWRVFSGNELGALLGWWLFTSWKEKNQDRSALKDTYMLSSTVSSKILRAIALKEGFHFEETLTGFKWMGNRAKQLIDQGKTVLFAFEEAIGYMCCPFVLDKDGVSAAVISAELASFLATKNLSLSQQLKAIYVEYGYHITKASYFICHDQETIKKLFENLRNYDGKNNYPKACGKFEISAIRDLTTGYDDSQPDKKAVLPTSKSSQMITFTFANGGVATMRTSGTEPKIKYYAELCAPPGNSDPEQLKKELNELVSAIEEHFFQPQKYNLQPKAD

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service